Order Entry
Belgium
Orders LinkContactUsLinkComponent
3,3',5,5'-Tetramethylbenzidine (TMB) QUICK, liquid substrate for ELISA

3,3',5,5'-Tetramethylbenzidine (TMB) QUICK, liquid substrate for ELISA

3,3',5,5'-Tetramethylbenzidine (TMB) QUICK, liquid substrate for ELISA
Certificates
3,3',5,5'-Tetramethylbenzidine (TMB) QUICK, liquid substrate for ELISA
3,3',5,5'-Tetramethylbenzidine (TMB) QUICK, liquid substrate for ELISA

2 Options Available Below

Product Details & Documents
Synonyms:  TMB, 3,3',5,5'-Tetramethyl-[1,1'-biphenyl]-4,4'-diamine
  • CAS Number:54827-17-7
  • Formula:C₁₆H₂₀N₂
  • MDL Number:MFCD00007748
  • Molecular Weight:240.35 g/mol
  • Storage Temperature:Refrigerator

Documents

Specifications for Product Family

Specification Test Results

  • Appearance:
    Clear colorless to yellow-green to light blue Solution
  • pH:
    3.50 to 3.90
  • Activity:
    90 to 110 %
  • Absorbance:
    ≤0.025 (450/620 nm, 0.2M H2SO4)
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle
    50 mL
    Log in for availability
    Log in for availability
    1 * 50 mL
    1 * 50 mL - 133,00 €
    133,00 €
    1 * 50 mL
       133,00 €

  • Glass bottle
    100 mL
    Log in for availability
    Log in for availability
    1 * 100 mL
    1 * 100 mL - 232,00 €
    232,00 €
    1 * 100 mL
       232,00 €

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.