0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Belgium
Orders Link
ContactUsLinkComponent
Buffers
pH Calibration Buffers
Buffer solution pH 4, citrate buffer
Buffer solution pH 4, citrate buffer
:
Thermo Fisher Scientific
Buffer solution pH 4, citrate buffer
:
Thermo Fisher Scientific
Buffer solution pH 4, citrate buffer
ACRO25858
Product Family Options
pH value
pH accuracy
pH buffer type
Colour
Pack type
Pk
ACRO258585000
4,00
±0,02
Citrate
Colourless
Plastic bottle
500 mL
1 * 500 mL
1 * 500 mL - 40,50 €
40,50 €
-
+
1 * 500 mL
40,50 €
-
+
ACRO258580010
4,00
±0,02
Citrate
Colourless
Plastic bottle
1 L
1 * 1 L
1 * 1 L - 60,60 €
60,60 €
-
+
1 * 1 L
60,60 €
-
+
ACRO258580050
4,00
±0,02
Citrate
Colourless
Plastic bottle
5 L
1 * 5 L
1 * 5 L - 128,00 €
128,00 €
-
+
1 * 5 L
128,00 €
-
+
Food and hygiene wiping paper, centrefeed roll, WypAll® Reach™
#: 115-2529
:
Wipes for delicate tasks, Kimtech™ Science
#: 115-2075
: