Caps, closed top
Supplier: EP SCIENTIFIC
À propos de cet article
PP, white.
- Very good chemical resistance
- Very good temperature resistance
Unprocessed caps are ideal for environmental sample bottles, jars, jugs and vials.
Certifications: Manufactured under an ISO certified quality management system
4 Options disponibles ci-dessous
Product Details & Documents
PP, white.
- Very good chemical resistance
- Very good temperature resistance
Unprocessed caps are ideal for environmental sample bottles, jars, jugs and vials.
Certifications: Manufactured under an ISO certified quality management system
Spécifications de la famille de produits
Spécifications
- Catalog No:
- EPSCB167-3300BL
- EPSCB167-3800BL
- EPSCB167-8900BL
- EPSCB167-1100BL
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement
Spécifications
Product Family Options
Informations sur le produit
- Cap SizeAvailabilityPrice
- EPSCB167-3300BLWith bonded PTFE Liner, bulkSpécifications :
Low Manufacturing Impact More
Specifications
Cat. No.EPSCB167-3300BLEnvironmentally PreferableLow Manufacturing ImpactEnvironmental certificationNAR Certificate of RetirementLow Manufacturing Impact - EPSCB167-3800BLWith bonded PTFE Liner, bulkSpécifications :
Low Manufacturing Impact More
Specifications
Cat. No.EPSCB167-3800BLEnvironmentally PreferableLow Manufacturing ImpactEnvironmental certificationNAR Certificate of RetirementLow Manufacturing Impact - EPSCB167-8900BLWith bonded PTFE Liner, bulkSpécifications :
Low Manufacturing Impact More
Specifications
Cat. No.EPSCB167-8900BLEnvironmentally PreferableLow Manufacturing ImpactEnvironmental certificationNAR Certificate of RetirementLow Manufacturing Impact - EPSCB167-1100BLWith bonded PTFE Liner, bulkSpécifications :
Low Manufacturing Impact More
Specifications
Cat. No.EPSCB167-1100BLEnvironmentally PreferableLow Manufacturing ImpactEnvironmental certificationNAR Certificate of RetirementLow Manufacturing Impact