Orders LinkContactUsLinkComponent
Corning® CoolCell® containers
Corning® CoolCell® containers
   Sustainable Options Available
Supplier:   Corning
Corning® CoolCell® containers
Supplier:  Corning
   Sustainable Options Available
Certificates
Corning® CoolCell® containers
13 Options Available Below
Product Details & Documents
CoolCell® alcohol-free cell freezing containers ensure standardised controlled-rate −1 °C/minute cell freezing in a −80 °C freezer, without alcohol or any fluids.
  • No alcohol. no fluids, no pre-cooling
  • No variability, all vials have uniform freeze rate
  • Radially symmetric design ensures vial consistency
  • No on-going cost, no alcohol purchase or disposal
  • No stuck lids, ergonomic lid comes off easily when frozen
  • Not cold to the touch when removing from the −80 °C freezer
  • Quick re-use time, ready to use again after five minutes
Proven for use with a variety of cell types including stem cells, primary cells, PBMC cell lines, insect cells, yeast and others. The patent-pending CoolCell® technology utilises a thermo-conductive alloy core and highly-insulative outer material to control the rate of heat removal and provide reproducible cell cryopreservation. CoolCell units are easy to use and deliver comparable results to expensive programmable freezers.
CoolCell® LX
For 12 standard 1,0 to 2,0 ml cryogenic vials, 1,0 ml fill per vial. Radially symmetric for uniform vial freezing. Numbered wells for easy sample identification. Beveled lid for secure gripping and easy opening. Exposed vial tops when lid is open for quick, organised removal of frozen samples.
CoolCell® FTS30
For 30 standard 1,0 to 2,0 ml cryogenic vials, 1,0 ml fill per vial. Controlled micro-convection for uniform freezing of 30 vials. Removable vial tray for one-step transfer of samples into and out of freezing chamber.
CoolCell® 5 mL LX
For 12 standard 3,5 to 5,0 ml fill cryogenic vials, 3,5 to 5,0 fill per vial. Radially symmetric for uniform vial freezing. Numbered wells for easy sample identification. Beveled lid for secure gripping and easy opening. Exposed vial tops when lid is open for quick, organised removal of frozen samples.
CoolCell® SV2 and CoolCell® SV10
For 12 standard 2,0 ml injectable ampules, 1,0 ml fill per ampule (SV2). For 6 standard 10,0 ml injectable ampules, 5,0 ml fill per ampule (SV10). Radially symmetric for uniform freezing of injectable ampules. Easy open lid. Exposed vial tops when lid is open for quick, organized removal of frozen samples.
CoolCell SV10 Stem Cell Cryopreservation System
includes the XT Starter Benchtop Cooler, Corning® CoolRack SV10 and CoolCell SV10 Cell Freezing Container for 6 injectable vials.
CoolCell® Filler Vials
To ensure cell freezing rate consistency and uniform results when using CoolCell containers, insert a CoolCell Filler Vial into empty wells when freezing less than a full load. Suitable for repeated use and compatible with CoolCell LX, CoolCell FTS30 and CoolCell 5 ml LX containers.
CoolCell® FTS30 Vial Module
FTS30 Vial Module is a holder for 30×1,0 ml or 2,0 ml cryogenic vials that allows one-step insertion and removal of all 30 vials at once. Fits into a standard 5,0×5,0×2,0" cryostorage box. Compatible with dry ice and liquid nitrogen.
Documents
Specifications for Product Family
Specifications
  • Catalog No:
  • 479-1840
  • 479-1841
  • 479-1842
  • 479-1843
  • 479-1844
  • CORN432138
  • 479-1848
  • 479-1849
  • 479-1850
  • 479-1851
  • 479-1845
  • 479-1918
  • 479-1919
  • Environmentally Preferable:
  • Safer Human and Environmental Health
  • Safer Human and Environmental Health
  • Safer Human and Environmental Health
  • Safer Human and Environmental Health
  • Safer Human and Environmental Health
  • Safer Human and Environmental Health
  • Safer Human and Environmental Health
  • Safer Human and Environmental Health
Specifications
Product Family Options
Product Information
  • For
    Colour
    Availability
    Price
  • 479-1840
    CoolCell®, fully covered tops
    CoolCell®, fully covered tops
    12 standard 1,0 mL to 2,0 mL cryogenic vials
    Purple
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 161,00 €
    161,00 €
    1 * 1 ST
       161,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    479-1840
    Environmentally Preferable
  • 479-1841
    CoolCell® LX, exposed tops
    CoolCell® LX, exposed tops
    12 standard 1,0 mL to 2,0 mL cryogenic vials
    Purple
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 202,00 €
    202,00 €
    1 * 1 ST
       202,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    479-1841
    Environmentally Preferable
  • 479-1842
    CoolCell® LX, exposed tops
    CoolCell® LX, exposed tops
    12 standard 1,0 mL to 2,0 mL cryogenic vials
    Green
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 202,00 €
    202,00 €
    1 * 1 ST
       202,00 €
    Specifications:
      Safer Human and Environmental Health
    More
    Specifications
    Cat. No.
    479-1842
    Environmentally Preferable
    Safer Human and Environmental Health
      Safer Human and Environmental Health
  • 479-1843
    CoolCell® LX, exposed tops
    CoolCell® LX, exposed tops
    12 standard 1,0 mL to 2,0 mL cryogenic vials
    Orange
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 202,00 €
    202,00 €
    1 * 1 ST
       202,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    479-1843
    Environmentally Preferable
  • 479-1844
    CoolCell® LX, exposed tops
    CoolCell® LX, exposed tops
    12 standard 1,0 mL to 2,0 mL cryogenic vials
    Pink
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 202,00 €
    202,00 €
    1 * 1 ST
       202,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    479-1844
    Environmentally Preferable
  • CORN432138
    CoolCell® LX, exposed tops
    CoolCell® LX, exposed tops
    12 standard 1,0 mL to 2,0 mL cryogenic vials (each one purple, orange, green and pink)
    Assorted colours
    Log in for availability
    Log in for availability
    1 * 4 ST
    1 * 4 ST - 685,00 €
    685,00 €
    1 * 4 ST
       685,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    CORN432138
    Environmentally Preferable
  • 479-1848
    CoolCell® FTS30, exposed tops
    CoolCell® FTS30, exposed tops
    30x1 ml or 2 ml cryogenic vials
    Purple
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 552,00 €
    552,00 €
    1 * 1 ST
       552,00 €
    Specifications:
      Safer Human and Environmental Health
    More
    Specifications
    Cat. No.
    479-1848
    Environmentally Preferable
    Safer Human and Environmental Health
      Safer Human and Environmental Health
  • 479-1849
    CoolCell® FTS30, exposed tops
    CoolCell® FTS30, exposed tops
    30x1 ml or 2 ml cryogenic vials
    Orange
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 552,00 €
    552,00 €
    1 * 1 ST
       552,00 €
    Specifications:
      Safer Human and Environmental Health
    More
    Specifications
    Cat. No.
    479-1849
    Environmentally Preferable
    Safer Human and Environmental Health
      Safer Human and Environmental Health
  • 479-1850
    CoolCell® FTS30, exposed tops
    CoolCell® FTS30, exposed tops
    30x1 ml or 2 ml cryogenic vials
    Green
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 552,00 €
    552,00 €
    1 * 1 ST
       552,00 €
    Specifications:
      Safer Human and Environmental Health
    More
    Specifications
    Cat. No.
    479-1850
    Environmentally Preferable
    Safer Human and Environmental Health
      Safer Human and Environmental Health
  • 479-1851
    CoolCell® FTS30, exposed tops
    CoolCell® FTS30, exposed tops
    30x1 ml or 2 ml cryogenic vials
    Pink
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 552,00 €
    552,00 €
    1 * 1 ST
       552,00 €
    Specifications:
      Safer Human and Environmental Health
    More
    Specifications
    Cat. No.
    479-1851
    Environmentally Preferable
    Safer Human and Environmental Health
      Safer Human and Environmental Health
  • 479-1845
    CoolCell® 5 ml LX, exposed tops
    CoolCell® 5 ml LX, exposed tops
    12x3,5 mL to 5 mL cryogenic vials
    Purple
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 271,00 €
    271,00 €
    1 * 1 ST
       271,00 €
    Specifications:
      Safer Human and Environmental Health
    More
    Specifications
    Cat. No.
    479-1845
    Environmentally Preferable
    Safer Human and Environmental Health
      Safer Human and Environmental Health
  • 479-1918
    CoolCell® SV2, exposed tops
    CoolCell® SV2, exposed tops
    12 standard 2.0 mL injectable ampules
    Purple
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 386,00 €
    386,00 €
    1 * 1 ST
       386,00 €
    Specifications:
      Safer Human and Environmental Health
    More
    Specifications
    Cat. No.
    479-1918
    Environmentally Preferable
    Safer Human and Environmental Health
      Safer Human and Environmental Health
  • 479-1919
    CoolCell® SV10, exposed tops
    CoolCell® SV10, exposed tops
    6 standard 10,0 mL injectable ampules
    Purple
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 386,00 €
    386,00 €
    1 * 1 ST
       386,00 €
    Specifications:
      Safer Human and Environmental Health
    More
    Specifications
    Cat. No.
    479-1919
    Environmentally Preferable
    Safer Human and Environmental Health
      Safer Human and Environmental Health
Accessories
Product Information
  • Catalog #
    For
    Availability
    Price
  • CoolCell® 2 mL filler vials
    CoolCell® 2 mL filler vials
    479-1961
    Use with CoolCell LX and CoolCell FTS30 containers
    Log in for availability
    1 * 6 ST - 29,20 €
Product Information
  • Catalog #
    Availability
    Price
  • Cryogenic vial grippers, assorted colours
    Cryogenic vial grippers, assorted colours
    479-1846
    Log in for availability
    1 * 5 ST - 56,00 €
Product Information
  • Catalog #
    For
    Availability
    Price
  • CoolCell® 5 mL filler vials
    CoolCell® 5 mL filler vials
    479-1962
    Use with CoolCell 5mL LX container
    Log in for availability
    1 * 6 ST - 29,20 €
Product Information
  • Catalog #
    For
    Availability
    Price
  • Removable cryogenic vial module, holds 30 cryogenic vials (1.0 mL or 2.0 mL)
    Removable cryogenic vial module, holds 30 cryogenic vials (1.0 mL or 2.0 mL)
    479-1852
    CoolCell FTS30
    Log in for availability
    1 * 10 ST - 121,00 €