Order Entry
Belgium
ContactUsLinkComponent
Heat resistant gloves, uvex profatherm XB40
Heat resistant gloves, uvex profatherm XB40
Catalog # 112-4536
Supplier:  UVEX
undefined
Heat resistant gloves, uvex profatherm XB40
Catalog # 112-4536
Supplier:  UVEX

Specifications

  • Glove size:
    11
  • Ambidextrous / hand-specific:
    Hand-specific
  • Disposable / reusable:
    Reusable
  • Colour:
    White
  • Length:
    400 mm
  • Model:
    XB40
  • EN norm:
    EN 388:2016 (2241B), EN 407 (X2XXXX)
  • Cuff style:
    Straight
  • Pk:
    6 PAIR

Specifications

About this item

This terry cotton safety glove is suitable for many areas of application, providing protection against heat (for contact heat up to 250 °C), cold and cuts.

  • Comfortable to wear
  • Good protection against cut injuries
  • Good temperature insulation

Ideal for use in foundries, in the steel industry, in plastics processing and in the metal processing industry.

Certifications: EN 388:2016 (2241B), EN 407 (X2XXXX)