ContactUsLinkComponent
Reverse osmosis water purification system, Barnstead™ Pacific™ RO
Reverse osmosis water purification system, Barnstead™ Pacific™ RO
Certificates
undefined
Reverse osmosis water purification system, Barnstead™ Pacific™ RO
Specifications
  • Catalog No:
  • BARN50132388
  • BARN50132386
  • BARN50132387
  • Model:
  • Pacific™ RO 20
  • Pacific™ RO 7
  • Pacific™ RO 12
  • Production flow rate:
  • 20 L/h
  • 7 L/h
  • 12 L/h
  • Bacteria:
  • 99%
  • 99%
  • 99%
  • W×D×H:
  • 372×330×603 mm
  • 372×330×603 mm
  • 372×330×603 mm
  • Weight:
  • 21 kg
  • 21 kg
  • 21 kg