Order Entry
Belgium
ContactUsLinkComponent
Foetal bovine serum, characterised, Australian origin, HyClone™
Foetal bovine serum, characterised, Australian origin, HyClone™
Certificates
undefined
Foetal bovine serum, characterised, Australian origin, HyClone™

Specifications

  • Catalog No:
  • SH30084.03-45
  • HYCLSH30084.03
  • HYCLSH30084.03HI
  • Origin:
  • Animal
  • Animal
  • Animal
  • Endotoxin:
  • ≤25 EU/ml
  • ≤25 EU/ml
  • ≤25 EU/ml
  • Haemoglobin:
  • ≤25 mg/dl
  • ≤25 mg/dl
  • ≤25 mg/dl
  • Storage temperature:
  • <–10 °C
  • <–10 °C
  • <–10 °C

Specifications

Frequently Bought Together