Orders LinkContactUsLinkComponent
VWR®, Density standards referenced at 40 °C
VWR®, Density standards referenced at 40 °C
Supplier:  VWR Chemicals
Certificates
VWR®, Density standards referenced at 40 °C
VWR®, Density standards referenced at 40 °C
8 Options Available Below
Product Details & Documents
These products can be used as calibration standards for density measurements by pycnometric, vibrational or hydrometer-based techniques.
  • High accuracy across an extensive density range (0,6752 - 3,0852 g/ml) at 40 °C
  • No toxic heavy metals used in any formulation
  • Can be used with any brand or type of density measuring instrument
  • Supplied in a high quality, tamper-proof, amber glass bottle
These standards are prepared gravimetrically (solute and solvent) using OIML traceable weights. The density values of the standards are established using fully calibrated reference pycnometers.
 : In accordance with ASTM D1480-12 for testing of density or relative density. A certificate of calibration is issued in accordance with the requirements of ISO/IEC 17025.
Product Family Options
Product Information
  • Density
    Reference temperature
    Pk
    Availability
    Price
  • Density standard
    0,7733 g/ml
    40 °C
    100 mL
    Log in for availability
    Log in for availability
    1 * 100 mL
    1 * 100 mL - 101,00 €
    101,00 €
    1 * 100 mL
       101,00 €
  • Density standard
    0,8945 g/ml
    40 °C
    100 mL
    Log in for availability
    Log in for availability
    1 * 100 mL
    1 * 100 mL - 101,00 €
    101,00 €
    1 * 100 mL
       101,00 €
  • Density standard
    0,9857 g/ml
    40 °C
    100 mL
    Log in for availability
    Log in for availability
    1 * 100 mL
    1 * 100 mL - 101,00 €
    101,00 €
    1 * 100 mL
       101,00 €
  • Density standard
    1,0152 g/ml
    40 °C
    100 mL
    Log in for availability
    Log in for availability
    1 * 100 mL
    1 * 100 mL - 237,00 €
    237,00 €
    1 * 100 mL
       237,00 €
  • Density standard
    1,0642 g/ml
    40 °C
    100 mL
    Log in for availability
    Log in for availability
    1 * 100 mL
    1 * 100 mL - 237,00 €
    237,00 €
    1 * 100 mL
       237,00 €
  • Density standard
    1,3217 g/ml
    40 °C
    100 mL
    Log in for availability
    Log in for availability
    1 * 100 mL
    1 * 100 mL - 237,00 €
    237,00 €
    1 * 100 mL
       237,00 €
  • Density standard
    1,5685 g/ml
    40 °C
    100 mL
    Log in for availability
    Log in for availability
    1 * 100 mL
    1 * 100 mL - 237,00 €
    237,00 €
    1 * 100 mL
       237,00 €
  • Density standard
    1,8994 g/ml
    40 °C
    100 mL
    Log in for availability
    Log in for availability
    1 * 100 mL
    1 * 100 mL - 237,00 €
    237,00 €
    1 * 100 mL
       237,00 €