ContactUsLinkComponent
Inclusion body solubilisation reagent, Pierce™
Inclusion body solubilisation reagent, Pierce™
Catalogus # PIER78115
CAS-nummer:  
undefined
Inclusion body solubilisation reagent, Pierce™
Catalogus # PIER78115
CAS-nummer:  
Specificaties
  • Pk:
    100 ml
  • Description:
    Inclusion Body Solubilisation Reagent
Specificaties
Over dit artikel
Inclusion Body Solubilisation Reagent is designed to retrieve expressed protein in soluble form after lysis and extraction with reagents.
  • Achieves the essential first step for subsequent insoluble protein refolding procedures
  • One easy to use solution
  • Proprietary denaturant can be easily removed by dialysis so that SDS-PAGE may be performed
  • Can accommodate additional components such as reducing agents or chelating agents