ContactUsLinkComponent
TCEP solution, neutral pH, Bond-Breaker™ Pierce™
TCEP solution, neutral pH, Bond-Breaker™ Pierce™
Catalog # PIER77720
CAS Number:  
undefined
TCEP solution, neutral pH, Bond-Breaker™ Pierce™
Catalog # PIER77720
CAS Number:  
Specifications
  • Pk:
    5 ml
  • Description:
    Bond-Breaker™ TCEP solution, neutral pH
Specifications
About this item
Bond-Breaker™ TCEP Solution (neutral pH) is a stable, 0,5 M solution of the thiol-free, phosphine-based TCEP compound, useful as a 10X stock for addition to SDS-PAGE sample loading buffers to reduce protein disulphide bonds.
  • Odourless
  • Selective and complete reduction of even the most stable water-soluble alkyl disulfides
  • Reduces protein disulphides at room temperature and pH 5 in less than five minutes
  • Resistant to air oxidation; nonvolatile and nonreactive toward other functional groups found in proteins