ContactUsLinkComponent
Antibody extender solution NC, Miser™
Antibody extender solution NC, Miser™
Catalog # PIER32110
CAS Number:  
undefined
Antibody extender solution NC, Miser™
Catalog # PIER32110
CAS Number:  
Specifications
  • Pk:
    500 ml
  • Description:
    Miser™ Antibody Extender Solution NC, sufficient solution for up to 20 mini blots or 1,600 cm2 nitrocellulose membrane
Specifications
About this item
Miser™ Antibody Extender Solution NC is a post-transfer nitrocellulose membrane-treatment reagent that enables Western blot detection with much less primary antibody.
  • Use at least three times less primary antibody than normal and retain comparable or better signal detectability
  • Easy to perform, 10-minute protocol
  • Works with both colorimetric and chemiluminescent detection modes and with HRP and AP systems