ContactUsLinkComponent
Test kits, oxygen cell test, Spectroquant®, Supelco®
Test kits, oxygen cell test, Spectroquant®, Supelco®
Catalog # 1.14694.0001
Supplier:  Merck
CAS Number:  
undefined
Test kits, oxygen cell test, Spectroquant®, Supelco®
Catalog # 1.14694.0001
Supplier:  Merck
CAS Number:  
Specifications
  • Chemical method:
    Winkler (modified)
  • Pk:
    25 Tests
  • Description:
    Oxygen cell test
  • M-Clarity quality level:
    MQ100
  • For unit:
    Prove, Nova 30/60, Move
  • Range:
    0,5 - 12,0 mg/l O₂
Specifications
About this item
All Spectroquant® test kits can be used with the Prove range of spectrophotometers and Nova 60/60A instruments. Tests can be used not only on photometers and spectrophotometer from Merck, but also on photometers and spectrophotometers from other suppliers (programming details available on request).
  • Very user friendly, validated methods
  • All the necessary reagents for the analysis are contained in the cells for most the test parameters
  • Packaging design protects the cells from fluctuation of temperature and moisture during storage and transport
  • The barcode on the cells ensures the correct method of analysis is chosen - eliminating mistakes and reducing set-up times
Note: The range quoted in the table below refers to the Prove and NOVA instruments. For some test kits the measuring range available for the Spectroquant® Multy and Move 100 instruments may be reduced.