Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

Glycerine base solution USP Test Solution (TS)

Supplier:   VWR Chemicals
Glycerine base solution USP Test Solution (TS)
Supplier:  VWR Chemicals
Certificates
Glycerine base solution USP Test Solution (TS)
Glycerine base solution USP Test Solution (TS)

2 Options Available Below

Product Details & Documents

Ready to use test solution (TS).

  • Produced according to Pharmacopoeia requirements
  • Reduces time and expenses
  • Independent, traceable and certified
  • Conforms to USP 34
  • Certificates of analysis and safety data sheets available
Synonyms:  1,2,3-Propanetriol, Propane-1,2,3-triol, Glycerol
  • CAS Number:56-81-5
  • Formula:HOCH₂CH(OH)CH₂OH
  • MDL Number:MFCD00004722
  • Molecular Weight:92.09 g/mol
  • Merck Index:13,04497
  • Melting Pt:18.6 °C
  • Density:1.2102 g/cm³ (20 °C)
  • Flash Pt:120 °C
  • Storage Temperature:Ambient
  • EINECS:0
  • UN:1824
  • ADR:8,II
Product Family Options

Product Information