Order Entry
Belgium
ContactUsLinkComponent
Ammonium chloride 10,5% USP Test Solution (TS)
Ammonium chloride 10,5% USP Test Solution (TS)
Catalog # 85329.180
Supplier:  VWR Chemicals
CAS Number:  
undefined
Ammonium chloride 10,5% USP Test Solution (TS)
Catalog # 85329.180
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • Seq:
    2440

Specifications

About this item

 :
 :53.49 g/mol
Merck Index:13,00510
Melting Pt:335 °C
Storage Temperature:Ambient
EINECS:0
CAS Number:12125-02-9
Formula:NH₄Cl
MDL Number:MFCD00011420