ContactUsLinkComponent
DAB substrate kit, metal enhanced, Pierce™
DAB substrate kit, metal enhanced, Pierce™
Catalog # PIER34065
CAS Number:  
undefined
DAB substrate kit, metal enhanced, Pierce™
Catalog # PIER34065
CAS Number:  
Specifications
  • Pk:
    250 ml
  • Description:
    Pierce™ Metal enhanced DAB substrate kit
Specifications
About this item
The Pierce™ Metal enhanced DAB substrate kit uses cobalt and nickel chloride in a special formulation of diaminobenzidine peroxidase substrate that yields intense colour for immunohistochemical staining.
  • Up to 50 times more sensitive than traditional DAB methods
  • Low background and high intensity
  • Only 2 components
  • Working solution stable for more than six hours at room temperature