Order Entry
Belgium
ContactUsLinkComponent
Pierce™ Firefly Signal Enhancer (100X)
Pierce™ Firefly Signal Enhancer (100X)
Catalog # PIER16180
CAS Number:  
undefined
Pierce™ Firefly Signal Enhancer (100X)
Catalog # PIER16180
CAS Number:  

Specifications

  • Pk:
    0,5 ml
  • Description:
    Pierce™ firefly signal enhancer, 100X
  • Storage conditions:
    Store in original container protected from direct sunlight in a dry, cool and well-ventilated area, but do not store above −20 °C
  • Detection method:
    Bioluminescence

Specifications

About this item

Pierce™ Firefly Signal Enhancer (100X) is designed to complement the Firefly Luciferase Flash and Glow assay and is recommended when maximum sensitivity is desired.

  • Up to 5-fold increase in Firefly Flash signal intensity
  • Up to 2-fold increase in Firefly Glow signal intensity and duration
  • Convenient and easy-to-use 100X solution