About this item
Test strips for the reflectometers RQflex® 10 plus and RQflex® 20.
- Fast analysis with reliable results
- Barcode calibration
- Low analysis costs
- Environmentally friendly
2 Options Available Below
Product Details & Documents
Test strips for the reflectometers RQflex® 10 plus and RQflex® 20.
- Fast analysis with reliable results
- Barcode calibration
- Low analysis costs
- Environmentally friendly
Documents
Specifications for Product Family
Specifications
Specifications
- Catalog No:
- 1.16978.0001
- 1.17942.0001
- M-Clarity quality level:
- MQ100
- MQ100
Specifications
Product Family Options
Product Information
- RangeChemical methodFor unitPkAvailabilityPrice
- 1.16978.0001Phosphate testPhosphate test5 - 120 mg/l PO₄Phosphormolybdenum blueRQflex® 2050 TestsLog in for availability1 * 50 Tests119,00 €Specifications:
More
Specifications
Cat. No.1.16978.0001M-Clarity quality levelMQ100 - 1.17942.0001Reflectoquant® 10 plus Phosphate testReflectoquant® 10 plus Phosphate test0,1 - 5,0 mg/l PO₄Phosphormolybdenum blueRQflex® 10 Plus100 TestsLog in for availability1 * 1 EA192,00 €Specifications:
More
Specifications
Cat. No.1.17942.0001M-Clarity quality levelMQ100
Complementary Products
Product Information
- Catalog #DescriptionModelAvailabilityPrice
- 1.17246.0001Reflectometer for test strips onlyRQflex 20Log in for availability1 * 1 ST - 1 420,00 €