ContactUsLinkComponent
Immunoassay microplates, biotin coated, 96-well strip plates, Pierce™
Immunoassay microplates, biotin coated, 96-well strip plates, Pierce™
Catalog # PIER15151
undefined
Immunoassay microplates, biotin coated, 96-well strip plates, Pierce™
Catalog # PIER15151
Specifications
  • Packed:
    5 plates per pack
  • Colour:
    Clear
  • Description:
    96-well strip microplate, 1×8 per strip
  • Pk:
    5
Specifications
About this item
Biotin coated plates can be used as the basis for a variety of biomolecular assay techniques requiring the immobilisation of probes that are conjugated with streptavidin, avidin, or other biotin-binding proteins.
  • Biotin group accessible for binding avidin or streptavidin
  • Pre-blocked to reduce non specific binding
  • Strip well plate format for the ultimate in convenience
  • Coat volume: 200 µl
  • Blocking volume: 300 µl