Order Entry
Belgium
ContactUsLinkComponent
Formic acid ≥90%, AnalaR NORMAPUR® ACS
Flammable
Corrosive
Formic acid ≥90%, AnalaR NORMAPUR® ACS
Catalog # 85495.290
Supplier:  VWR Chemicals
CAS Number:  
undefined
Flammable
Corrosive
Formic acid ≥90%, AnalaR NORMAPUR® ACS
Catalog # 85495.290
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

Specification Test Results

  • Assay:
    89.0 - 91.0 %
  • Dilution test:
    Passes test ACS
  • SO3 (Sulphite):
    Passes test
  • Colouration:
    Max. 10 APHA
  • Acetic acid:
    Max. 0.04 %
  • Evaporation residue:
    Max. 20 ppm
  • Heavy metals (by ICP-MS):
    Max. 5 ppm
  • Cl (Chloride):
    Max. 10 ppm
  • NH4 (Ammonium):
    Max. 50 ppm
  • SO4 (Sulphate):
    Max. 20 ppm
  • Fe (Iron):
    Max. 5 ppm
  • Conforms to ACS:
    Passes test

Specification Test Results

About this item

 :
 :46.03 g/mol
Merck Index:13,04265
Density:1.208 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:0
UN:1779
ADR:8 (3),II
 :GHS02
 :GHS05
CAS Number:64-18-6
Formula:HCO₂H
MDL Number:MFCD00003297