ContactUsLinkComponent
Immunoglobulin purification kit, Thiophilic adsorption kit, Pierce™
Immunoglobulin purification kit, Thiophilic adsorption kit, Pierce™
Catalog # PIER44916
CAS Number:  
undefined
Immunoglobulin purification kit, Thiophilic adsorption kit, Pierce™
Catalog # PIER44916
CAS Number:  
Specifications
  • Pk:
    1 KIT
  • Description:
    Thiophilic adsorption kit, 4 columns, sufficient for purifying up to 60 mg immunoglobulin per column
  • Seq:
    01
Specifications
About this item
Pierce™ Thiophilic adsorbent is a protein-free immunosorbent resin for gentle preferential binding and purification of immunoglobulins from a variety of sources including mouse, rat, rabbit, goat and human. This Pierce™ Thiophilic Adsorption Kit is a purification kit containing columns pre-packed with the resin and ready-to-use buffers.
  • High capacity - gently binds and purifies
  • Broad antibody specificity
  • Simple and fast - quickly fractionate and purify the immunoglobulin component
  • Binding condition is non denaturing and the elution buffer yields concentrated, salt-free immunoglobulin
Thiophilic adsorption is based on the ability of some proteins, particularly immunoglobulins, to bind to an immobilised ligand that contains a sulfone group in close proximity to a thioether. Salts that interact with water molecules, such as potassium and ammonium sulfate, promote this binding by driving the protein into close proximity with the ligand.