Orders LinkContactUsLinkComponent
Test Strips, Peroxide
Test Strips, Peroxide
Test Strips, Peroxide
Supplier:  MACHEREY-NAGEL
Certificates
Test Strips, Peroxide
3 Options Available Below
Product Details & Documents
QUANTOFIX Peroxide test strips enables the quick and reliable determination of peroxides in water–based liquids. The simple handling makes the test strip particularly user–friendly: simply dip it into the sample, wait briefly and read off the result – in less than a minute.
  • Peroxide - Redox reaction (white → blue or brown for 705-1154)
The test strip can be evaluated with the included colour scale as well as instrumental with the QUANTOFIX Relax strip reader. In addition to a convenient measuring procedure, this also offers automated and digital documentation of the results. QUANTOFIX Peroxide is ideal for the food industry. The test strips can be used to check rinsing and washing processes of equipment quickly, easily and reliably – disinfectant residues in bottles and yoghurt pots can be easily identified. The same applies to medical devices. QUANTOFIX Peroxide  is an ideal companion for food quality and patient safety.
Specifications for Product Family
Specifications
Specifications
  • Catalog No:
  • MANA91319
  • 705-0313
  • 705-1154
  • Method:
  • Peroxide - Redox reaction (white → blue)
  • Peroxide - Redox reaction (white → blue)
  • Peroxide - Redox reaction (white → brown)
  • Parameters:
  • All reagents included
  • All reagents included
  • All reagents included
  • Compliance:
  • No hazardous material
  • No hazardous material
  • No hazardous material
  • Contents:
  • 100 test strips in a tube, instruction leaflet
  • 100 test strips in a tube, instruction leaflet
  • 100 test strips in a tube, instruction leaflet
  • Instrument:
  • QUANTOFIX Relax
  • QUANTOFIX Relax
  • QUANTOFIX Relax
  • Platform:
  • Semi-quantitative test strips
  • Semi-quantitative test strips
  • Semi-quantitative test strips
  • Shelf life (months):
  • 30 (from production)
  • 30 (from production)
  • 30 (from production)
  • Weight:
  • 43,5 g
  • 49 g
  • 43,1 g
Specifications
Product Family Options
Product Information
  • Range
    Chemical method
    Dimensions
    Tests per kit
    Pk
    Availability
    Price
  • MANA91319
    QUANTOFIX® peroxide 25
    QUANTOFIX® peroxide 25
    0 • 0,5 • 2 • 5 • 10 • 25 - mg/L - H₂O₂; 0,5 - 25 - mg/L - H₂O₂
    Redox reaction (peroxidase/redox indicator)
    40×40×120 mm (packaging)
    100 Tests
    100 Tests
    Log in for availability
    Log in for availability
    1 * 1 EA
    1 * 1 EA - 32,30 €
    32,30 €
    1 * 1 EA
       32,30 €
    Specifications:
    More
    Specifications
    Cat. No.
    MANA91319
    Method
    Peroxide - Redox reaction (white → blue)
    Parameters
    All reagents included
    Compliance
    No hazardous material
    Contents
    100 test strips in a tube, instruction leaflet
    Instrument
    QUANTOFIX Relax
    Platform
    Semi-quantitative test strips
    Shelf life (months)
    30 (from production)
    Weight
    43,5 g
  • 705-0313
    QUANTOFIX® peroxide 100
    QUANTOFIX® peroxide 100
    0 • 1 • 3 • 10 • 30 • 100 - mg/L - H₂O₂; 1 - 100 - mg/L - H₂O₂
    Redox reaction (peroxidase/redox indicator)
    43×126×43 mm (packaging)
    100 Tests
    100 Tests
    Log in for availability
    Log in for availability
    1 * 1 EA
    1 * 1 EA - 40,38 €
    40,38 €
    1 * 1 EA
       40,38 €
    Specifications:
    More
    Specifications
    Cat. No.
    705-0313
    Method
    Peroxide - Redox reaction (white → blue)
    Parameters
    All reagents included
    Compliance
    No hazardous material
    Contents
    100 test strips in a tube, instruction leaflet
    Instrument
    QUANTOFIX Relax
    Platform
    Semi-quantitative test strips
    Shelf life (months)
    30 (from production)
    Weight
    49 g
  • 705-1154
    QUANTOFIX® peroxide 1000
    QUANTOFIX® peroxide 1000
    0 • 50 • 150 • 300 • 500 • 800 • 1000 - mg/L - H₂O₂; 50 - 1000 - mg/L - H₂O₂
    Redox reaction (peroxidase/redox indicator)
    40×40×120 mm (packaging)
    100 Tests
    100 Tests
    Log in for availability
    Log in for availability
    1 * 1 EA
    1 * 1 EA - 33,20 €
    33,20 €
    1 * 1 EA
       33,20 €
    Specifications:
    More
    Specifications
    Cat. No.
    705-1154
    Method
    Peroxide - Redox reaction (white → brown)
    Parameters
    All reagents included
    Compliance
    No hazardous material
    Contents
    100 test strips in a tube, instruction leaflet
    Instrument
    QUANTOFIX Relax
    Platform
    Semi-quantitative test strips
    Shelf life (months)
    30 (from production)
    Weight
    43,1 g