Order Entry
Belgium
ContactUsLinkComponent
Octyl-β-D-thioglucopyranoside non-ionic detergent, Pierce™
Octyl-β-D-thioglucopyranoside non-ionic detergent, Pierce™
Catalog # PIER28351
CAS Number:  
undefined
Octyl-β-D-thioglucopyranoside non-ionic detergent, Pierce™
Catalog # PIER28351
CAS Number:  

Specifications

  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :308.44 g/mol
CAS Number:85618-21-9
Formula:C₁₄H₂₈O₅S
MDL Number:MFCD00012189