Order Entry
Belgium
Orders LinkContactUsLinkComponent
Blocker™ Blocking Buffer, BLOTTO in TBS

Blocker™ Blocking Buffer, BLOTTO in TBS

Blocker™ BLOTTO in TBS, blocking buffer, 1 L
Catalog # PIER37530
Blocker™ Blocking Buffer, BLOTTO in TBS
Catalog # PIER37530
CAS Number:  
Blocker™ Blocking Buffer, BLOTTO in TBS
Blocker™ Blocking Buffer, BLOTTO in TBS

Blocker™ BLOTTO in TBS, blocking buffer, 1 L

CAS Number:  
Price
197,00 €
Log in for availability
1 * 1 L
Product Details & Documents

Blocker™ BLOTTO is a ready-to-use 5% solution of non fat powdered milk in Tris-buffered saline for Western blot, ELISA, IHC and other detection methods.

  • Convenient as can be diluted as needed
  • Easy to use, formulated with anti-foaming agent and thimerosal-free preservative
  • Flexible may be used for multiple applications

Caution: For research use only. Not for use in diagnostic procedures.

Specifications

Specifications

  • Pk:
    1 L
  • Description:
    Blocker™ BLOTTO in TBS, blocking buffer

Specifications