About this item
Pierce™ Maleimide activated plates are ideal for binding sulfhydryl-containing molecules that are difficult to coat onto polystyrene plates, such as peptides that contain a terminal cysteine. Available in clear for colorimetric assays, white for chemiluminescent assays and black for fluorescent assays.
- Pre-blocked bovine serum albumin (BSA) to reduce non specific binding
- Binding capacity is 100 to 150 pmol of a sulfhydryl-containing peptide (307 kDa) per well
- Activation level: 100 µl
Pierce™ Maleimide activated plates provide covalent attachment (plate capture) of sulfhydryl-containing molecules, such as cysteine-terminated peptides. These coated plates are an especially useful tool for assessing specific anti-hapten antibody titers during antibody production.
Maleimide activated plates are available in clear for colorimetric assays, white for chemiluminescent assays and black for fluorescent assays. Highlights Spontaneous immobilization of cysteine-containin peptides and proteins with free sulfhydryls Pre-blocked to reduce nonspecific bindingFor Research Use Only. Not for use in diagnostic procedures.