Orders LinkContactUsLinkComponent
HyClone™ CDM4HEK293
Canonical URL fetch failed:ERROR
HyClone™ CDM4HEK293
HyClone™ CDM4HEK293
Certificates
HyClone™ CDM4HEK293
HyClone™ CDM4HEK293
2 Options Available Below
Product Details & Documents
HyClone™ CDM4HEK293 media are chemically-defined, animal-derived component free (ADCF) cell culture media.
  • Allows for direct or sequential adaptation
  • Component traceability
  • Manufactured under cGMP
  • Metabolically designed for high cell yield and recombinant protein/vector production
HyClone CDM4HEK293 is a regulatory-friendly medium developed through the Metabolic Pathway Design to support the extremely high productivity and cell density in suspension cultures.
Product Family Options
Product Information
  • Media Format
    Pk
    Availability
    Price
  • SH30858.02
    CDM4HEK293 medium without phenol red, L-glutamine, HEPES
    CDM4HEK293 medium without phenol red, L-glutamine, HEPES
    Liquid
    1 L
    Log in for availability
    Log in for availability
    1 * 1 L
    1 * 1 L - 100,00 €
    100,00 €
    1 * 1 L
       100,00 €
  • HYCLSH30859.01
    CDM4HEK293 medium without phenol red, L-glutamine, HEPES, sodium bicarbonate
    CDM4HEK293 medium without phenol red, L-glutamine, HEPES, sodium bicarbonate
    Powder
    5 L
    Log in for availability
    Log in for availability
    1 * 5 L
    1 * 5 L - 325,00 €
    325,00 €
    1 * 5 L
       325,00 €
Recommandations
Les recommandations seront personnalisées en fonction de vos préférences d’achat uniquement si vous avez donné votre consentement en activant l’option " Améliorer mon expérience d’achat " sur la page "Mon compte".
Dans le cas contraire, vous recevrez des recommandations génériques.