ContactUsLinkComponent
Freund's incomplete adjuvant, Imject™
Freund's incomplete adjuvant, Imject™
Catalog # PIER77145
CAS Number:  
undefined
Freund's incomplete adjuvant, Imject™
Catalog # PIER77145
CAS Number:  
Specifications
  • Pk:
    1 SET
  • Description:
    Imject™ Freund's incomplete adjuvant, 5×10 ml
  • Environmentally Preferable:
Specifications
About this item
Imject™ Freund's Incomplete Adjuvant (FIA) is a popular non specific stimulator of the immune response for use in antibody production immunisation protocols.
  • Enhances immune response in booster injections
  • Typically used for subsequent boosts after initial injection with FCA
  • Water-in-oil emulsion without Mycobacterium
Delivery: Supplied ready to use in 10 ml ampoules.