Visual colour kits, carbonate hardness, MQuant®, Supelco®
Supplier: Merck
Certificates
About this item
Quick tests, visual colour comparison.
- Results in approximately 5 minutes
- Easy to use
- Application-oriented measuring ranges
- Portable
- A range of economical refill packs (RP) are available
MQuant® titrimetric and colorimetric methods:
This range comprises both titrimetric and colorimetric tests for medium concentrations. With its broad range of available test parameters, these quick tests are the method of choice for aquaria, fish farming, aquatic ecology, environmental, cooling and boiler water analysis.
** 1 drop
Delivery: Each test kit contains all the necessary reagents, reaction cuvettes, colour comparison chart and extensive instructions for use.
Caution: ** 1 drop
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together