Acetonitrile, anhydrous (max. 0.001 % H₂O) ≥99.95% for DNA synthesis
:75-05-8
:
:H₃CCN
:MFCD00001878
:41.05 g/mol
:13,00071
:81.6 °C (1013 hPa)
:–45.7 °C
:0.782 g/cm³ (20 °C)
:200-835-2
:1648
:3,II
:Filtered through a 0.2 µm filter, packaged under nitrogen.
:5 °C (closed cup)
:01-2119471307-38
:GHS02
:GHS07
:Ambient
- Environmentally Preferable:Safer Human and Environmental Health
- Environmental certification:Chem21 Solvent Selection Guide recommended by default and after discussion
- Assay:Min. 99.95 %
- IR Spectrum:Passes test
- Acidity:Max. 0.001 meq/g
- Alkalinity:Max. 0.0002 meq/g
- Colouration:Max. 10 APHA
- Evaporation residue:Max. 1 ppm
- Water:Max. 10 ppm
Frequently Bought Together