ContactUsLinkComponent
Dulbecco's modified eagle medium limiting media (DMEM-LM)
Dulbecco's modified eagle medium limiting media (DMEM-LM)
Catalog # PIER30030
CAS Number:  
Dulbecco's modified eagle medium limiting media (DMEM-LM)
Catalog # PIER30030
CAS Number:  
Specifications
  • Media Format:
    Liquid
  • Pk:
    500 ml
  • Description:
    DMEM (-)L-leucine, (-)L-methionine with 4,5g/l glucose, 4,0 mM L-glutamine, sodium pyruvate and phenol red