Order Entry
Belgium
ContactUsLinkComponent
HyClone™ IMDM, Cell culture media, powdered
HyClone™ IMDM, Cell culture media, powdered
Catalog # HYCLSH30005.02
CAS Number:  
undefined
HyClone™ IMDM, Cell culture media, powdered
Catalog # HYCLSH30005.02
CAS Number:  

Specifications

  • Media Format:
    Powder
  • Pk:
    10 L
  • Description:
    IMDM, with L-glutamine and HEPES, without alpha-thioglycerol, 2×5 l
  • Seq:
    01
  • Size:
    10 L

Specifications

About this item

Dry powdered media for supporting cell culture growth and biomanufacturing processes.