Order Entry
Belgium
ContactUsLinkComponent
Pierce™ Recombinant Protein G, Biotinylated
Pierce™ Recombinant Protein G, Biotinylated
Catalog # PIER29988
CAS Number:  
undefined
Pierce™ Recombinant Protein G, Biotinylated
Catalog # PIER29988
CAS Number:  

Specifications

  • Pk:
    0,5 mg
  • Description:
    Recombinant protein G, biotinylated

Specifications

About this item

Biotinylated recombinant Protein G can be used to probe and detect mouse and human antibodies, especially IgG isotypes, in western blotting, ELISA, IHC and other immunoassay protocols.

  • Contains two Fc-binding domains per protein
  • Does not bind human IgM, IgD or IgA
  • Efficient than Protein A for mouse (IgG1), human (IgG3) and rat, goat and cow antibodies
  • Less efficient than Protein A for guinea pig, pig, dog and cat