Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

Lithium nitrate, ionisation buffer solution, Specpure®

Lithium Nitrate Buffer Solution, 100 mL
Catalog # 45293.AE
Lithium nitrate, ionisation buffer solution, Specpure®
Catalog # 45293.AE
CAS Number:  
Lithium nitrate, ionisation buffer solution, Specpure®
Lithium nitrate, ionisation buffer solution, Specpure®

Lithium Nitrate Buffer Solution, 100 mL

CAS Number:  
Price
120,00 €
Log in for availability
1 * 100 mL
Product Details & Documents

CAS No.: 7790-69-4
Molecular Formula: 1% Li (as Li₂CO₃) in 5% HNO₃
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00011094

Specifications

Specifications

  • Pk:
    100 mL
  • Description:
    Lithium Nitrate Buffer Solution
  • UN No.:
    3264
  • Cat. No.:
    45293.AE
  • Molecular formula:
    1% Li (as Li2CO3) in 5% HNO3
  • Physical form:
    Liquid
  • MDL No.:
    MFCD00011094
  • CAS No.:
    7790-69-4

Specifications