Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

Sodium Cacodylate, 0,1 M Buffer solution, pH 6,5

Sodium cacodylate buffer soln, 0,1 M, 6,5, 100 mL
  J60367.AE
Sodium Cacodylate, 0,1 M Buffer solution, pH 6,5
  J60367.AE
 :  
Sodium Cacodylate, 0,1 M Buffer solution, pH 6,5
Sodium Cacodylate, 0,1 M Buffer solution, pH 6,5

Sodium cacodylate buffer soln, 0,1 M, 6,5, 100 mL

 :  
 
148,00 €
   
1 * 100 mL
 

Liquid.

CAS No.: 6131-99-3
Storage Temperature: 2 to 8 °C
Physical Form: Liquid
MDL No.: MFCD00149079

 

 

  • Concentration:
    0,1 M
  • Pk:
    100 mL
  • Description:
    Sodium cacodylate buffer soln
  • pH value:
    6,5
  • Cat. No.:
    J60367.AE
  • Storage temperature:
    2°C to 8°C
  • Physical form:
    Liquid
  • MDL No.:
    MFCD00149079
  • CAS No.:
    6131-99-3