Order Entry
Belgium
Orders LinkContactUsLinkComponent
Temperature Sensor, T201, radiometer analytical

Temperature Sensor, T201, radiometer analytical

Temperature sensor with 1 m cable and cinch plug, −10…+105 °C, 7,5×103 mm
Catalog # HACHE51M001
Supplier:   Hach
Temperature Sensor, T201, radiometer analytical
Catalog # HACHE51M001
Supplier:  Hach
Temperature Sensor, T201, radiometer analytical
Temperature Sensor, T201, radiometer analytical

Temperature sensor with 1 m cable and cinch plug, −10…+105 °C, 7,5×103 mm

Price
447,00 €
Log in for availability
1 * 1 ST
Product Details & Documents

This temperature sensor features a glass body, fixed cable and 10 kohm resistance thermistor. The T201's cinch (RCA) connector compatible with radiometer PHM2xx, CDM2xx, PHM9x, IONxxx and TIMxxx meters.

  • Meets the analysis needs
  • Available with a variety of lengths, diameters and reference systems

 : Supplied with radiometer analytical T201 temperature sensor with certificate of conformity.

Documents

Specifications
  • Range:
    −10…+105 °C
  • Description:
    Temperature sensor with 1 m cable and cinch plug
  • Ø×L:
    7,5×103 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.