Orders LinkContactUsLinkComponent
pH electrode, Radiometer Analytical
pH electrode, Radiometer Analytical
pH electrode, PHG301-9 (sensor only, external reference probe needed), Laboratory pH electrode, 0 - 12, 0...80 °C, Glass, 12×103 mm, Screw cap (no cable), General purposes
Catalog # HACHE11M001
Supplier:   Hach
pH electrode, Radiometer Analytical
Catalog # HACHE11M001
Supplier:  Hach
pH electrode, Radiometer Analytical
pH electrode, Radiometer Analytical
pH electrode, PHG301-9 (sensor only, external reference probe needed), Laboratory pH electrode, 0 - 12, 0...80 °C, Glass, 12×103 mm, Screw cap (no cable), General purposes
Price
250,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
These glass electrodes are compatible with Hach sensION+ benchtop pH meters (or any meter with a BNC connection). The PHG301 pH electrode should be used with Ag/AgCl or calomel reference electrodes.
  • Made with highly shock-resistant glass, making them extremely tough
  • Average lifetime of a correctly maintained Radiometer glass pH electrode is approximately two years
The PHG301 is a screw cap electrode and requires the CL114 cable for connection to a meter.
Delivery information: Supplied with certificate of conformity.
Specifications
  • Connection:
    Screw cap (no cable)
  • Ø×L:
    12×103 mm
  • For:
    General purposes
  • Type:
    Laboratory pH electrode
  • Description:
    pH electrode, PHG301-9 (sensor only, external reference probe needed)
  • Material:
    Glass
  • Temp.:
    0...80 °C
  • pH:
    0 - 12