Transfer buffer, Tris-glycine,10X, AccuGENE™
Catalog # 733-1629
Supplier: LONZA
CAS Number:
Specifications
- Pk:1 L
- Description:AccuGENE™ 10X Tris-glycine buffer, 0,25 M Tris base, 1,92 M glycine
- Environmentally Preferable:
Specifications
About this item
AccuGENE™ electrophoresis buffers are ready-to-use solutions ideal for a wide range of molecular biology applications.
- Ready to use solutions
- Reliable manufactured according to strict quality control standards to ensure lot to lot consistency
- Guaranteed DNase-, RNase- and protease-free
Each reagent is prepared with 18 megOhm water, filtered using a 0,2 micron filter, and filled into sterile bottles.