ContactUsLinkComponent
Hybridisation buffer, SSPE 20X, AccuGENE™
Hybridisation buffer, SSPE 20X, AccuGENE™
Catalog # 733-1638
Supplier:  LONZA
CAS Number:  
Hybridisation buffer, SSPE 20X, AccuGENE™
Catalog # 733-1638
Supplier:  LONZA
CAS Number:  
Specifications
  • Pk:
    1 L
  • Description:
    AccuGENE™ 20X SSPE buffer, 3,0 M NaCl, 0,2 M NaH₂PO₄, H₂O, 0,02 M EDTA, pH 7,4
Specifications
About this item
AccuGENE™ electrophoresis buffers are ready-to-use solutions ideal for a wide range of molecular biology applications.
  • Ready to use solutions
  • Reliable manufactured according to strict quality control standards to ensure lot to lot consistency
  • Guaranteed DNase-, RNase- and protease-free
Each reagent is prepared with 18 megOhm water, filtered using a 0,2 micron filter, and filled into sterile bottles.