Order Entry
Belgium
ContactUsLinkComponent
Digital heating shaking block heater
Digital heating shaking block heater
  BARN88880028
undefined
Digital heating shaking block heater
  BARN88880028

 

  • No. of blocks:
    1
  • Orbit:
    2 mm
  • Speed range:
    150 - 1500 min⁻¹ (96-well block);
    150 - 1200 min⁻¹ (0,5, 1,5 and 50 ml tube blocks)
  • Temperature control range:
    Ambient +5...100 °C
  • Description:
    Digital heating shaking block heater, 230 V
  • Model:
    Digital shaking block heater
  • Temperature accuracy:
    ±0,5 °C @ 80 °C
  • Weight:
    8,3 kg
  • W×D×H:
    76×334×170 mm
  • Pk:
    1

 

 

This can be used as a drybath, shaker or both. The wide temperature range covers a broad range of applications including thawing, incubation, enzyme reactions, blood urea nitrogen determinations, and immunoassays.

  • PID control ensures consistent shaking results and precise temperature control
  • Stores ten programs and up to ten steps per program
  • Interval shaking function allows shaking and nonshaking in same program
  • Cord set with various plug types
  • Low-profile design
  • Easily changed blocks in multiple sizes for maximum flexibility
 : Blocks must be ordered separately.