Order Entry
Belgium
ContactUsLinkComponent
 
Digital heating/cooling block heater
  BARN88880030
undefined
Digital heating/cooling block heater
  BARN88880030

 

  • Accuracy:
    0,55 °C @ 37 °C
  • No. of blocks:
    1
  • Description:
    Digital heating cooling block heater, 230 V
  • Model:
    digital heating/cooling block heater
  • Temperature setting:
    +4...+95 °C
  • Weight:
    5 kg
  • W×D×H:
    276×334×165 mm
  • Pk:
    1
  • Temp. stability:
    ±0,15 °C

 

 

Compact programmable heating/cooling block heater with digital display can be used for all common applications in laboratory. Applications include enzyme reactions, transformation, DNA extractions, reactions that need to processed at or near room temperature and warming and cooling of reagents.

  • PID control ensures consistent precise temperature control
  • Stores 10 programs and 10 steps per program
  • Cord set with various plug types
  • Low-profile design
  • Blocks easily interchanged
 : Blocks must be ordered separately.

Blocks