ContactUsLinkComponent
Electrode filling solution, IntelliCAL™ PHC281
Electrode filling solution, IntelliCAL™ PHC281
Catalog # HACH2965026
Supplier:  Hach
CAS Number:  
undefined
Electrode filling solution, IntelliCAL™ PHC281
Catalog # HACH2965026
Supplier:  Hach
CAS Number:  
Specifications
  • Pk:
    59 mL
  • Description:
    Internal filling solution for use with PHC281 IntelliCAL™ Ultra pH probes, 2,44M KCl, 59 ml bottle with dispenser cap