Microinjectors, Nanoject III™
Nanoject III™, 100 - 240 V for US, UK, Europe and Australia
Catalog # DRUM3-000-207
Supplier: DRUMMOND
Jump to:
Product Details & Documents
The Nanoject III™ microinjector is designed to perform single manual injections and multiple injection cycle recipes. The auto homing feature eliminates the need to rehome the unit and reduce jamming of the plunger.
- User-friendly programmable touchscreen operation
- Accurate injection volumes down to 0,6 nanoliters
- Ensures accurate and consistent injection volumes with hydraulic technology
- O-rings not required to secure the micropipette
- Lower injection volume capability
Delivery information: Supplied with 4 interchangeable adapter plugs.
Specifications
- Description:Nanoject III™, 100 - 240 V for US, UK, Europe and Australia
Accessories
Product Information
- Catalog #AvailabilityPrice
- Replacement wire plunger
Product Information
- Catalog #AvailabilityPrice
- Replacement collet kit with chuck, green seal, wire plunger, and allan wrenchReplacement collet kit with chuck, green seal, wire plunger, and allan wrenchDRUM3-000-030-KLog in for availability1 * 1 KIT - 196,00 €
Product Information
- Catalog #AvailabilityPrice
- Footswitch with remote capability for fill, empty and inject functionsFootswitch with remote capability for fill, empty and inject functionsDRUM3-000-032Log in for availability1 * 1 ST - 765,00 €
Product Information
- Catalog #AvailabilityPrice
- Silicon seal, green