NMR pipettes, KIMBLE®
NMR pipette, with extended tip (229 mm)
SCERSP897085-0009
Manufactured from 51 expansion borosilicate glass conforming to USP type I and ASTM E438, type I, class B requirements.
These extended tip NMR pipettes are for retrieving and adding samples in NMR tubes.
- Description:NMR pipette, with extended tip (229 mm)
- Seq:02