Vitamin B12 (cyanocobalamin) FCC
Supplier: Spectrum Chemical
Certificates
About this item
CAS Number:68-19-9
:
Formula:C₆₃H₈₈CoN₁₄O₁₄P
MDL Number:MFCD00151092
:1355.38 g/mol
Merck Index:12,10152
Melting Pt:320 °C
Storage Temperature:Refrigerator
EINECS:200-680-0
Specification Test Results
- Assay:96% Min.
Specification Test Results
Frequently Bought Together