Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Belgium
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Standards
Environmental Analysis Standards
Dissolved oxygen standard solution
Dissolved oxygen standard solution
Dissolved oxygen standard solution
HACH40149
:
Hach
:
Dissolved oxygen standard solution
HACH40149
:
Hach
:
Pk:
500 mL
Description:
Iodate-Iodide standard solution, 0.00125 N equivalent 10 mg/L DO