Order Entry
Belgium
ContactUsLinkComponent
 
Hardness standard solution, APA6000
Catalog # HACH2058016
Supplier:  Hach
CAS Number:  
undefined
Hardness standard solution, APA6000
Catalog # HACH2058016
Supplier:  Hach
CAS Number:  

Specifications

  • Concentration:
    0,50 mg/l as CaCO3
  • Pk:
    946 mL
  • Description:
    Hardness standard solution

Specifications

About this item

Hardness standard solution, 0,50 mg/l as CaCO₃ traceable to NIST.