Orders LinkContactUsLinkComponent
Tablet count kits for water hardness
Tablet count kits for water hardness
Total hardness test, 250 tests, 250
Catalog # PALIAD-1016
Supplier:   Palintest
Tablet count kits for water hardness
Catalog # PALIAD-1016
Supplier:  Palintest
CAS Number:  
Tablet count kits for water hardness
Tablet count kits for water hardness
Total hardness test, 250 tests, 250
CAS Number:  
Price
514,00 €
Log in for availability
1 * 250 ST
Product Details & Documents
Tablet Count kits provide a simple method of analysis for an important range of water parameters. The tests are particularly useful for the routine control of industrial waters such as boiler waters and cooling water systems, as well as swimming pools, natural waters and similar applications.
  • Cost effective and provides quick results on-site
  • Can be used as a stand alone kit or combined with comparator or photometer
  • Wide range of test concentrations can be measured
Specifications
Specifications
  • Pk:
    250
  • Description:
    Total hardness test, 250 tests
Specifications