Order Entry
Belgium
Orders LinkContactUsLinkComponent
Vanillin NF

Vanillin NF

Irritant
Supplier:   Spectrum Chemical
Vanillin NF
Supplier:  Spectrum Chemical
Certificates
Vanillin NF

3 Options Available Below

Product Details & Documents
Synonyms:  4-Hydroxy-3-methoxybenzaldehyde
  • CAS Number:121-33-5
  • Formula:C₈H₈O₃
  • MDL Number:MFCD00006942
  • Molecular Weight:152.15 g/mol
  • Merck Index:13,09998
  • Boiling Pt:285 °C (1013 hPa)
  • Melting Pt:∼82 °C
  • Density:1.056 g/cm³ (20 °C)
  • Flash Pt:153 °C
  • Storage Temperature:Ambient
  • EINECS:204-465-2
Specifications for Product Family

Specification Test Results

  • Assay:
    97% Min.
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Poly pail
    2,5 kg
    Log in for availability
    Log in for availability
    1 * 2,5 kg
    1 * 2,5 kg - 836,00 €
    836,00 €
    1 * 2,5 kg
       836,00 €

  • Fiber drum
    12 kg
    Log in for availability
    Log in for availability
    1 * 12 kg
    1 * 12 kg - 2 590,00 €
    2 590,00 €
    1 * 12 kg
       2 590,00 €

  • 50 kg
    Log in for availability
    Log in for availability
    1 * 50 kg
    1 * 50 kg

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.