Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Belgium
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Brij® 35
Brij® O2
Brij® O2
Brij® O2
Catalog # SPCMB1676-200LTBL
Supplier:
Spectrum Chemical
CAS Number:
Expand image
Brij® O2
Catalog # SPCMB1676-200LTBL
Supplier:
Spectrum Chemical
CAS Number:
Specifications
Pk:
200 L
Seq:
5549
Specifications
About this item
Product Details
:
EINECS:
500-016-2
CAS Number:
9004-98-2
MDL Number:
MFCD00074369
Product Details