Diammonium hydrogen orthophosphate, granular NF
- Assay:96% Min.
Frequently Bought Together
Acetone ≥99%, TECHNICAL
: VWR Chemicals
Sodium chloride regeneration salt, TECHNICAL for water softening
: VWR Chemicals
Potassium chloride ≥99%, GPR RECTAPUR®
: VWR Chemicals
Magnesium sulphate heptahydrate USP, Ph. Eur., BP
: VWR Chemicals