Order Entry
Belgium
Orders LinkContactUsLinkComponent
D-(-)-Fructose, granular USP

D-(-)-Fructose, granular USP

Poly pail, 12 kg
Catalog # SPCMFR106-12KG
Supplier:   Spectrum Chemical
D-(-)-Fructose, granular USP
Catalog # SPCMFR106-12KG
Supplier:  Spectrum Chemical
CAS Number:  57-48-7
D-(-)-Fructose, granular USP
D-(-)-Fructose, granular USP

Poly pail, 12 kg

CAS Number:  57-48-7
Price
1 360,00 €
Log in for availability
1 * 12 kg
Product Details & Documents

Form: Granular

Synonyms:  D(-)-Levulose, (3S,4R,5R)-1,3,4,5,6-pentahydroxyhexan-2-one, β-D-(-)-Fructose, Fructose
  • CAS Number:57-48-7
  • Formula:C₆H₁₂O₆
  • MDL Number:MFCD00148910
  • Molecular Weight:180.16 g/mol
  • Merck Index:13,04295
  • Boiling Pt:552 °C (1013 hPa)
  • Melting Pt:100…110 °C
  • Density:1.59 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:200-333-3
Specifications

Specification Test Results

  • Assay:
    98% Min.

Specifications

  • Pack type:
    Poly pail
  • Pk:
    12 kg

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.